Learn More
Bellco Glass, Inc. 10mL Short Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile
SDP
Supplier:  Bellco Glass, Inc. 229211B00
Encompass_Preferred
Compact design of short serological pipet provides ergonomic benefit and reduces storage spaceShorter length helps prevent touching pipet tip to surfaces in the hoodIdeal for students and those new to pipettingAll serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identification Pack Description50 Individually Wrapped/BagCase Quantity200SterileYesNon-PyrogenicYesPrimary ColorClearSecondary ColorOrangePrimary MaterialPolystyreneMaximum Volume10mLGraduatedYesGraduation Increments (Subdivisions)2/10mLRNase/DNase FreeYes
Catalog No. 50-233-0672