Learn More
Bellco Glass, Inc. 10mL Wide Tip Serological Pipet, Individual Paper/Plastic Wrapper Packed in Bags, Sterile
SDP
Supplier:  Bellco Glass, Inc. 229212B00
Encompass_Preferred
All serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identificationPrinted indicator arrow for quickly locating negative graduations when using a pipet controller10mL Wide Tip Serological Pipet for viscous solutions – Wide Tip opening is 4x larger than standard 10mL pipet Pack Description50 Individually Wrapped/BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorOrangePrimary MaterialPolystyreneMaximum Volume10mLGraduatedYesGraduation Increments (Subdivisions)1/10mLRNase/DNase FreeYes
Catalog No. 50-233-0674