Learn More
Pacer Global LLC 10mL Serological Pipet, Individually Wrapped, Paper/Plastic, Carton, Sterile, 200 per case
SDP
Supplier:  Pacer Global LLC MLSSPI10J
Encompass_Preferred
10 mL in 1/10mL Serological Pipets have clearly marked ascending and descending graduations to accurately measure the amount of liquid that is aspirated and dispensed. Certified DNase free, RNase free, non-pyrogenic, and non-cytotoxic. Meet the requirements of BSE/TSE. Sterilized by Gamma irradiation SAL10 (ISO11137). Each serological pipet is manufactured in a class 100,000 clean room environment, under ISO13485 and ISO9001 quality management system. Additional info on MLS-SPI-10J: Not Autoclavable. Temperature Range: -20°C to +50°C. Graduation in 1/10mL. Latex Free. Shelf Life of 3 years after month of production. Negative graduations up to -0.3 ml (total volume =13 ml). Printed with a batch number for traceability. Pipet made of GPPS (General polystyrene), Compliance with USP Class VI. Each pipet comes with a filter plug. Filter made of PO (Polyolefin). Orange Color Coding. Clear Color. Individually wrapped in peel-to-open wrap. Dispenser box for easy opening 50/pack,200/case.
Catalog No. NC1931447