Learn More
Biopioneer 10 mL Serological Pipet, 500/pk
SDP
Supplier:  Biopioneer GEX0100S01500PK
Encompass_Preferred
10ml Serological Pipet, Individual Wrapped (one side paper, one side plastic), Sterilized pipet can be used for variety of purpose, 500/pk. BioPioneer Serological pipettes are sterilized by Gamma irradiation, certified Non-pyrogenic, DNase/RNase Free.
Catalog No. NC1890999