Learn More
Ace Glass, Inc. 10mm PTFE inner bearing, straight adapter style
SDP
Supplier:  Ace Glass, Inc. 806606
Encompass_Preferred
10mm self-aligning, lubricant-free PTFE Trubore bearing, straight adapter style. Use with ACE polished stainless steel or glass 10mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.
Catalog No. 50-955-8169