Learn More
Chemglass Life Sciences Stirring Shaft, 10mm, Polished, 530mm OAL
SDP
Supplier:  Chemglass Life Sciences CG2075B04
Encompass_Preferred
Stirring Shaft, 10mm, Polished, 530mm OAL. A 10mm precision ground and polished stirrer shaft for use with PTFE bearings and CG-2091 and CG-2092 PTFE agitators. Flat ground section extends half way up shaft and permits positive seating of the PTFE locking screws.
Catalog No. CG2075B04