Learn More
Cayman Chemical 10Nitrooleated17
SDP
Supplier:  Cayman Chemical 10558100 UG
Encompass_Preferred
An internal standard for the quantification of arachidonic acid by GC- or LC-MS
Catalog No. 50-288-8900