Learn More
Abcam 10nm Gold Nanoparticles (1 OD), 100ML
Supplier:  Abcam AB269933100ML
Encompass_Preferred
10nm Gold Nanoparticles (1 OD) (ab269933) is an aqueous suspension of spherical metallic nanoparticles manufactured to the highest standard to ensure: Uniform spherical shape, Ultra high quality, Batch to batch consistency and High efficiency antibody binding for maximum sensitivity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-2815