Learn More
USA Scientific Inc 10-place upright freezer rack
Supplier:  USA Scientific Inc 26022500
Encompass
10-place (2 long x 5 high) upright freezer rack for 2 in H boxes, 10 15/16 in L x 5 1/2 in W x 11 in H, stainless steel
Catalog No. 50-274-4746