Learn More
Trajan Scientific 10UL FIXED NEEDLE SYR, 5CM NDL
Supplier:  Trajan Scientific 002005
Encompass_Preferred
10uL fixed needle syringe with 5 cm 0.47 mm OD cone tipped needle
Catalog No. NC2184263