Learn More
Bellco Glass, Inc. 10ul Inoculating Loop Indivual
SDP
Supplier:  Bellco Glass, Inc. 229612000
Encompass_Preferred
DescriptionFlexible for easy access to small volume tubes and dishesColor coded for easy identificationGentle on delicate agar or other surfaces Pack Description1/WrapCase Quantity600SterileYesNon-pyrogenicYesPrimary ColorBluePrimary MaterialPolypropyleneWorking Volume10μLGraduatedNoRNase/DNase FreeYes
Catalog No. 50-233-0736