Learn More
Ace Glass, Inc. 10 microliter gas-tight, fixed needle syringe
SDP
Supplier:  Ace Glass, Inc. 593101
Encompass
10 microliter gas-tight, fixed needle syringe. Syringe has a PTFE coated plunger with a precision PTFE tip for a leak-tight seal. Designed for highest performance in such applications as liquid or gas chromatography, handling of corrosive gases and liquids, radioactive materials and sterile solutions. Accuracy and reproducibility of plus or minus 1%.
Catalog No. 50-978-615