Learn More
Chemglass Life Sciences Chemglass Extra-Long 10 μL Pipette Tips, Clear, Non-Sterile, Bulk Nest - For Single and Multichannel Pipettors
SDP
Supplier:  Chemglass Life Sciences CLS3499E010
Encompass_Preferred
Chemglass extra-long 10 μL clear pipette tips offer precise low-volume transfer and extended reach for sampling from deep or narrow vessels. Bulk nest packaging simplifies storage and reduces handling for high-throughput workflows.
  • Extra-long design for extended reach and access.
  • 10 μL nominal volume for low-volume dispensing.
  • Ultra-clear polypropylene construction for solvent resistance and visibility.
  • Non-sterile, bulk nest packaging (1,000/pack; 10,000/case).
  • Certified RNase-free, DNase-free, human DNA-free, and pyrogen-free.
  • Low-retention, fine-point geometry reduces sample adhesion.
  • Universal fit for most single and multichannel pipettors.
Catalog No. 50-273-3212