Learn More
Trajan Scientific Hamilton 10l Syringe, PAL3 FN, 22s, PTFE, Pt. 3
Supplier:  Trajan Scientific HAM.207813
Encompass
Hamilton CTC C-Line PAL3 Syringe Gastight 10µL, OD 6.6 mm, Scale 60mm, Fixed Needle Gauge 22s, PointStyle 3, (LC applications)
Catalog No. NC1122670