Learn More
Quality Lab Accessories 10UM DISTEK FILTER 1000PK
Supplier:  Quality Lab Accessories FIL010DK
Encompass
10 Micron Filter Discs, UHMW Polyethylene, Distek compatible (Jar/1000)
Catalog No. NC9750293