Learn More
Diagnostic Biosystems 10X CITRATE BUFFER PH 70 10X Citrate Buffer For Heat Induced Epitope Recovery, pH 7.0
Supplier:  Diagnostic Biosystems K036
Encompass_Preferred
10X Citrate Buffer For Heat Induced Epitope Recovery, pH 7.0 is designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin embedded tissue sections. The use of this buffer in combination with heat (often by microwave, water bath, or pressure cooker) has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue. This buffer is supplied as a 10X stock solution.
Catalog No. 50-240-7381