Learn More
LI COR BIOTECH LLC 10X Orange Loading Dye
Supplier: LI COR BIOTECH LLC 92710100
10X Orange Loading Dye is used as a loading buffer for DNA gel electrophoresis. This loading dye is not visualized when imaged on Odyssey Imagers. The primary application for 10X Orange Loading Dye is for electrophoretic mobility shift assays (EMSA). When run on a 5% native acrylamide 1X TBE gel, the orange dye migrates with the 20-28 bp DNA fragment.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.