Learn More
Boston Bioproducts Inc 10X TBS 2000ML
Supplier:  Boston Bioproducts Inc BM3012L
Encompass_Preferred
TRIS-Buffered Saline (TBS, 10X) For Western Blot Washing For western blot washing, Tris 0.5M, NaCl 1.5M, pH 7.4 +-.01 1L
Catalog No. NC0203600