Learn More
Teknova 10X TBS with Tween-20 Kit, 1000mL, Sterile. (1M Tris, 1.5M NaCl, 0.5% Tween) 8 pack.
SDP
Supplier:  Teknova T9501PK
Encompass
10X TBS with Tween-20 Kit, 1000mL, Sterile. (1M Tris, 1.5M NaCl, 0.5% Tween) 8 pack.
Catalog No. 50-227-4086