Learn More
Uline 10X8X8IN COURRUGATED BOX 25PK
Supplier:  Uline S17994
Encompass_Preferred
DIMENSIONS:Outside: 10 3/8 x 8 3/8 x 8 5/8" (+/- 1/8")All FedEx and all UPS Dimensional Weight: 6 lbsMATERIAL: C fluteFEATURES:All boxes open along the length (first dimension).Seams are glued.SPECIFICATIONS:Meets ASTM D5118TERMINOLOGY:200# Test vs 32 ECT (Edge Crush Test):200# Test:60% stronger bursting strength.Recommended by UPS for shipping up to 40 lbs.32 ECT:Recommended by UPS for shipping up to 30 lbs.Pound Test Refers to Bursting Strength:200# test boxes withstand 200 lbs/sq.in. of side wall pressure before bursting.ECT Refers to Stacking Strength:32 ECT boxes withstand 32 lbs/linear inch of stacking weight before crushing.An alternative for lightweight shipments.BOX STRENGTH GUIDELINES: (Maximum weight per box)FIBRE BOX HANDBOOK UPS GUIDELINES32 ECT Single Wall 65 lbs 30 lbs200# Single Wall 65 lbs 40 lbs275# Single Wall 95 lbs 65 lbs350# Single Wall 120 lbs 80 lbs200# Double Wall 80 lbs 60 lbs275# D
Catalog No. NC4041790