Learn More
Medchemexpress LLC 10Z-Hymenialdisine | 82005-12-7 | 500 UG
SDP
Supplier:  Medchemexpress LLC HYN6794500UG
Encompass_Preferred
10Z-Hymenialdisine ((Z)-Hymenialdisine) is a natural bioactive pyrrole alkaloid and a pan kinase inhibitor, demonstrating anticancer activities. It is intended for research use only and not for sale to patients.
  • Natural bioactive pyrrole alkaloid
  • Pan kinase inhibitor
  • Exhibits anticancer activities
  • Displays moderate effect on ARK5, VEGFR-2, SAK, and PDGFR protein kinases
  • Potent inhibition of RAF/MEK-1/MAPK cascade with an IC50 value of 6 nM
Catalog No. 50-004-17780