Learn More
Medchemexpress LLC (±)11(12)-EET-d11 | 2699607-19-5 | 98.0% | 10 UG
SDP
Supplier:  Medchemexpress LLC HY130494S10UG
Encompass_Preferred
(±)11(12)-EET-d11 is a deuterium labeled form of (±)11(12)-EET, functioning as an NLRP3 inflammasome inhibitor. It is used in research for anti-inflammatory, angiogenic, and cardioprotective effects. This compound can serve as a tracer and an internal standard for quantitative analysis by techniques like NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Inhibits NLRP3 inflammasome
  • Used in anti-inflammatory, angiogenic, and cardioprotective research
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
Catalog No. 50-003-39788