Learn More
Medchemexpress LLC 11,14,17-eicosatrienoic acid, (11Z,14Z,17Z)- | 17046-59-2 | 99.3% | 306.48 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY108398B10MG
Encompass_Preferred
11(Z),14(Z),17(Z)-Eicosatrienoic acid is an unsaturated fatty acid that can maintain the continued replication of functional mitochondria in *Saccharomyces cerevisiae* (KD115).
  • Maintains the continued replication of functional mitochondria in *Saccharomyces cerevisiae* (KD115).
Catalog No. 50-003-47505