Learn More
Medchemexpress LLC 11,15-O-dimethyl-PGE2 | 211230-67-0 | 98.3% | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1218425MG
Encompass_Preferred
ONO-AE 248 (CAS 211230-67-0) is a selective prostaglandin EP3 receptor agonist used in research to investigate cardioprotective mechanisms. It has been shown to reduce myocardial infarct size in animal models and is used in pharmacology and mechanistic studies involving protein kinase C and K(ATP) channels.
  • Selective EP3 receptor agonist for pharmacology studies.
  • Demonstrated cardioprotective activity in myocardial infarction models.
  • High purity suitable for research applications.
  • Molecular formula C22H36O5; molecular weight ~380.5 g/mol.
  • Supplied in a 25 MG quantity for laboratory use.
  • Store as recommended to preserve stability and activity.
Catalog No. 50-002-31207