Learn More
Medchemexpress LLC (11-((4-hydroxy-3-methoxybenzoyl)oxy)undecyl)triphenylphosphonium bromide | 1254044-38-6 | 99.8% | C37H44BrO4P | 10MG
SDP
Supplier:  Medchemexpress LLC HY13586910MG
Encompass_Preferred
Mito-apocynin (C11) is a mitochondria-targeted derivative of apocynin in which the apocynin moiety is conjugated to a triphenylphosphonium cation. It is supplied as a high-purity research reagent intended for preclinical studies of mitochondrial oxidative stress, neuroprotection, and related mitochondrial metabolism pathways.
  • Targets mitochondria via a triphenylphosphonium moiety.
  • Provides antioxidant activity to mitigate mitochondrial oxidative stress.
  • Suitable for in vitro and in vivo preclinical research applications.
  • High purity, approximately 99.8%.
  • Available in milligram-scale quantities for laboratory use.
Catalog No. 50-002-59849