Learn More
Trajan Scientific 11.5 mm OD BTO premium high-temp center guide GC septa PK25
SDP
Supplier:  Trajan Scientific 041899
Encompass_Preferred
11.5 mm OD BTO premium high-temp center guide GC septa PK25
Catalog No. 50-272-5277