Learn More
Medchemexpress LLC HY-114464 5mg Medchemexpress, 11-Beta-hydroxyandrostenedione CAS:382-44-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1144645MG
Encompass_Preferred
Medchemexpress, HY-114464 5mg 11-Beta-hydroxyandrostenedione CAS:382-44-5 11-Beta-hydroxyandrostenedione (4-Androsten-11β-ol-3,17-dione) is a steroid mainly found in the the adrenal origin (11β-hydroxylase is present in adrenal tissue, but absent in ovarian tissue). 11-Beta-hydroxyandrostenedione is a 11β-hydroxysteroid dehydrogenase (11βHSD) isozymes inhibitor. As 4-androstenedione increases, measuring plasma 11-Beta-hydroxyandrostenedione can distinguish the adrenal or ovarian origin of hyperandrogenism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9361