Learn More
Medchemexpress LLC 11-bromoundecan-1-ol | 1611-56-9 | MFCD00004752 | 251.21 g/mol | C11H23BrO | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01390750G
Encompass_Preferred
11-Bromoundecan-1-ol is an ω-brominated primary alcohol used as a synthetic intermediate and PROTAC linker for constructing bifunctional degraders and other small molecules. It is a C11 alkyl bromide with formula C11H23BrO and molecular weight 251.21 g/mol, intended for research and medicinal chemistry applications.
  • Useful as a PROTAC linker and linker-building block.
  • Primary alcohol and alkyl bromide functional groups enable substitution and coupling reactions.
  • Molecular formula C11H23BrO; molecular weight 251.21 g/mol.
  • Stable at room temperature for typical storage; store in solvent at low temperature for extended shelf life.
  • Supplied in gram-scale packages suitable for synthesis and small-scale research.
Catalog No. 50-003-02287