Learn More
Medchemexpress LLC 11-cis-Vaccenyl acetate | 6186-98-7 | 99.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12890010MG
Encompass_Preferred
11-cis-Vaccenyl acetate is a male-specific lipid that mediates aggregation behavior in both male and female flies. It activates olfactory neurons located in T1 sensilla on the antenna of both male and female flies.
  • Male-specific lipid
  • Mediates aggregation behavior in flies
  • Activates olfactory neurons in T1 sensilla
Catalog No. 50-003-40292