Learn More
Enzo Life Sciences 11-dehydro-TXB2 ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900092
Encompass_Preferred
The 11-dehydro-TXB2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit offers ultrasensitive quantification of 11-d-TXB2 in a variety of sample matrices with negligible reactivity with related molecules in a convenient 96-well format offers higher throughput analysis compared to conventional analytical methods. Alternative name: 11-dehydro-Thromboxane B2. Sensitivity: 4.31 pg/ml (range 9.8 - 10,000 pg/ml). Use/Stability: Store all components at +4°C, except standard and conjugate at -20°C. Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent . Content: GxR IgG Microtiter plate, Assay buffer, Conjugate, Antibody, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Catalog No. 50-200-6839