Learn More
Cayman Chemical 11-DehydrocortIcosteron 5mg
SDP
Supplier:  Cayman Chemical 329075 MG
Encompass_Preferred
An endogenous mineralocorticoid; increases Na+/K+-ATPase mRNA expression in vascular smooth muscle cells and inhibits aldosterone action in B. marinus toad bladder tissue in a concentration-dependent manner; decreases the sodium/creatine ratio and increases the potassium/creatine ratio in rat urine in a dose-dependent manner in a model of carbenoxolone-induced 11β-hydroxysteroid dehydrogenase inhibition; chronic administration increases plasma glucocorticoids levels, body weight gain, and adiposity, as well as induces insulin resistance in mice
Catalog No. 50-295-0852