Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11β-HSD1-IN-8 | 386704-15-0 | MFCD01862654 | 95.0% | 247.17 | C10H8F3NO3 | 250 MG
SDP

Supplier:  Medchemexpress LLC HYW129359250MG

Encompass_Preferred

11β-HSD1-IN-8 is a small-molecule inhibitor of 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) with a reported IC50 of 2.3 μM for the human enzyme. Supplied as a high-purity solid research reagent, it is intended for in vitro studies of metabolic and cognitive disorders such as diabetes and cognitive decline.

  • Selective inhibition of 11β-HSD1 (IC50 2.3 μM).
  • Intended for in vitro research of metabolic and cognitive disorders.
  • High stated purity of 95.0%.
  • Solid form; off-white to light yellow appearance.
  • Storage: powder stable at -20°C (3 years) or 4°C (2 years).
  • Molecular formula C10H8F3NO3; molecular weight 247.17.

Catalog No. 50-002-83251


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.