Learn More
Medchemexpress LLC 11-Keto-beta-boswellic acid | 17019-92-0 | 470.68 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN20561MG
Encompass_Preferred
11-Keto-beta-boswellic acid is a pentacyclic triterpenic acid derived from the oleogum resin of the Boswellia serrata tree, commonly known as Indian Frankincense. It demonstrates anti-inflammatory activity primarily by inhibiting 5-lipoxygenase (5-LOX) and subsequent leukotriene and nuclear factor-kappa B (NF-κB) activation.
  • Derived from Boswellia serrata (Indian Frankincense)
  • Exhibits anti-inflammatory activity
  • Inhibits 5-lipoxygenase (5-LOX)
  • Inhibits nuclear factor-kappa B (NF-κB) activation
  • For research use only
Catalog No. 50-004-15239