Learn More
Medchemexpress LLC 11(α)-Methoxysaikosaponin F | 104109-37-7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN42151MG
Encompass_Preferred
11(α)-Methoxysaikosaponin F is a triterpenoid saponin isolated from Bupleurum marginatum Wall.ex DC(ZYCH). It is considered a promising therapeutic for liver fibrosis.
  • Isolated from Bupleurum marginatum Wall.ex DC(ZYCH).
  • Promising therapeutic for liver fibrosis.
  • Exhibits an IC50 of 387.7 nM with viability of hepatic stellate cells-T6 (HSCs-T6).
  • Known for numerous targets, important network positions, and strong inhibitory activity.
  • Applicable in COVID-19 immunoregulation.
Catalog No. 50-004-18968