Learn More
Medchemexpress LLC 11-octadecenoic acid, (11E)- | 693-72-1 | MFCD00002734 | 98.0% | 282.46 g·mol⁻¹ | C18H34O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1134275MG
Encompass_Preferred
trans-Vaccenic acid is a naturally occurring trans fatty acid (11-octadecenoic acid, (11E)-) provided as an analytical standard for research and analytical applications. It is used in studies of lipid metabolism, fatty acid synthesis, and as a reference standard for chromatographic and mass spectrometric analysis.
  • High purity: 98.0% suitable for analytical and research applications.
  • Available in small quantities (5 mg) for standard preparation and method development.
  • Useful as a reference standard for lipid profiling and chromatographic calibration.
  • Precursor to conjugated linoleic acid formation in biological tissues.
  • Supplied with a certificate of analysis for batch-specific quality data.
Catalog No. 50-002-63547