Learn More
Medchemexpress LLC 11-oxo-mogroside V | 126105-11-1 | 1285.42 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN050110MM/1ML
Encompass_Preferred
11-oxo-mogroside V is a natural sweetener that exhibits strong antioxidant activity. It significantly inhibits reactive oxygen species (O2-, H2O2, and *OH) with EC50 values of 4.79, 16.52, and 146.17 μg/mL, respectively.
  • Acts as a natural sweetener.
  • Exhibits strong antioxidant activity.
  • Significantly inhibits reactive oxygen species.
  • Offers inhibitory effects on O2-, H2O2, and *OH.
Catalog No. 50-003-89979