Learn More
Medchemexpress LLC HY-N0501 1mg Medchemexpress, 11-oxo-mogroside V CAS:126105-11-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN05011MG
Encompass_Preferred
Medchemexpress, HY-N0501 1mg 11-oxo-mogroside V CAS:126105-11-1 11-oxo-mogroside V is a natural sweetener, isolated from the fruits of Momordica grosvenori, exhibits strong antioxidant activity. It exhibits significant inhibitory effects on reactive oxygen species ( O 2 - , H 2 O 2 and *OH ) with EC 50 of 4.79, 16.52, and 146.17 μg/mL, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3597