Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11-Oxomogroside IIa | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN704110MG

Encompass_Preferred

11-Oxomogroside IIa is a cucurbitane glycoside extracted from the fruits of *Siraitia grosvenorii*. It demonstrates inhibitory effects against Epstein-Barr virus early antigen (EBV-EA) activation and exhibits weak inhibitory effects on the activation of NOR 1, a nitric oxide (NO) donor. This product is for research use only.

  • Purity of 99.92%
  • Molecular weight of 799.00
  • Chemical formula C42H70O14
  • Appears as a white to light yellow solid
  • Classified as a terpenoid and triterpene
  • Derived from *Siraitia grosvenorii* plants
  • Inhibits Epstein-Barr virus early antigen activation
  • Weakly inhibits nitric oxide donor activation

Catalog No. 50-004-18993


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.