Learn More
Cayman Chemical 11-trns LeukotrIen D4 100ug
SDP
Supplier:  Cayman Chemical 20330100 UG
Encompass_Preferred
11-trans LTD4 is a C-11 double bond isomer of LTD4. LTD4 undergoes slow temperature-dependent isomerization to 11-trans LTD4 during storage. 11-trans LTD4 retains about 10-25 % of the potency for contraction of guinea pig ileum, trachea and parenchyma compared to LTD4.{11179,11181} It exhibits an ED50 ranging between 12-60 nM for contraction of guinea pig trachea.{11181,11178}
Catalog No. 50-298-2489