Learn More
Cayman Chemical 11-trns LeukotrIen E4 100ug
SDP
Supplier:  Cayman Chemical 20430100 UG
Encompass_Preferred
Slow isomerization of the C-11 double bond of LTE4 leads to the formation of 11-trans LTE4.{589} 11-trans LTE4 is equipotent to LTE4 in contracting guinea pig ileum.{2236}
Catalog No. 50-298-2732