Learn More
Cayman Chemical 11 Z 14 Z 17 ZEIcosatrIen 10mg
SDP
Supplier:  Cayman Chemical 9019210 MG
Encompass_Preferred
Eicosatrienoic Acid (20:3ω-3) is a rare PUFA of the ω-3 series. In normal humans, it represents less than 0.25% of serum phospholipid fatty acids. However, it is one of the most active essential fatty acids when assayed for the inhibition of fatty acid elongation/desaturation reactions which convert dietary C-18 fatty acids to C-20 eicosanoid precursors.{8460}
Catalog No. 50-289-6081