Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 11(Z),14(Z)-Eicosadienoic acid | 5598-38-9 | ≥98.0% | 308.50 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY149589100MG

Encompass_Preferred

11(Z),14(Z)-Eicosadienoic acid is an unsaturated fatty acid that inhibits [3H] leukotriene B4 (LTB4) binding to pig neutrophil membranes with a Ki of 3 μM. It also possesses inflammatory activities.

  • Unsaturated fatty acid
  • Inhibits [3H] leukotriene B4 (LTB4) binding
  • Possesses inflammatory activities
  • Purity: ≥98.0%
  • Molecular weight: 308.50
  • Appearance: Liquid, colorless to light yellow
  • Solubility in DMSO: 125 mg/mL
  • Solubility in Ethanol: ≥ 100 mg/mL

Catalog No. 50-003-59461


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.