Learn More
Apexbio Technology LLC 11(Z)-Eicosenoic Acid 5561-99-9 250mg
SDP
Supplier:  Apexbio Technology LLC C4128250
Encompass_Preferred
11(Z)-Eicosenoic Acid (CAS 5561-99-9) is a monounsaturated long-chain fatty acid featuring a 20-carbon aliphatic chain with a cis double bond at the 11 position While its precise molecular targets are not fully delineated long-chain fatty acids of this type are known to participate in cell membrane biosynthesis lipid signaling and modulation of metabolic pathways 11(Z)-Eicosenoic Acid is commonly utilized in research to study lipid metabolism membrane fluidity and as a model compound in investigations of fatty acid synthesis and elongation Its structural characteristics make it valuable for examining the physiological effects of unsaturated fatty acids in cellular and metabolic research contexts
Catalog No. 50-000-91258