Learn More
Cayman Chemical 11 ZHexadecenyl2ArachIdo 5mg
SDP
Supplier:  Cayman Chemical 341825 MG
Encompass_Preferred
A plasmalogen; serum levels are decreased in dogs with myxomatous mitral valve disease fed a diet including medium chain triglycerides, fish oil, carnitine precursors, magnesium, and antioxidants
Catalog No. 50-294-8305