Learn More
Sino Biological Influenza A H3N2 (A/Brisbane/10/2007) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11056V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Brisbane/10/2007(H3N2)) hemagglutinin (ABW23353.1) (Met1-Trp530), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6149