Learn More
eMolecules 110782-31-5 | 2'-OMe-A(Bz) Phosphoramidite | ChemScene | MFCD00792622 | 887.975 | C48H54N7O8P | 97.000 | CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c2ccc(OC)cc2)c2ccc(OC)cc2)O[C@H]1n1cnc2c(NC(=O)c3ccccc3)ncnc12 | 10g | 687111568
Supplier: eMolecules 687111568
2'-OMe-A(Bz) Phosphoramidite | ChemScene | 110782-31-5 | MFCD00792622 | 887.975 | C48H54N7O8P | 97.000 | CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c2ccc(OC)cc2)c2ccc(OC)cc2)O[C@H]1n1cnc2c(NC(=O)c3ccccc3)ncnc12 | 10g | 687111568