Learn More
Sino Biological Recombinant Human R-Spondin 1/RSPO1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11083H08H100
Encompass_Preferred
A DNA sequence encoding the human RSPO1 (NP_001033722.1) (Met 1-Ala 263) was expressed fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6500