Learn More
Chemrus Inc 110ML FILTER FUNNEL 10 MICRON
Supplier:  Chemrus Inc CR1008110
Encompass
Chemrus 110 mL polypropylene barrel-shaped disposable filter funnel, 10 micron pore size polyethylene fritted disc
Catalog No. 50-823-07