Learn More
Sino Biological Recombinant Human DNMT2 Protein (GST Tag)
Supplier:  Sino Biological 11224H09B100
Encompass_Preferred
A DNA sequence encoding the human TRDMT1 isoform a (NP_004403.1) (Met 1-Glu 391) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-6710