Learn More
Sino Biological Recombinant Human IGSF3 Protein (His Tag)
Supplier:  Sino Biological 11290H08H20
Encompass_Preferred
A DNA sequence encoding the human IGSF3 isoform 1 (O75054-1) extracellular domain (Met 1-Ala 1124) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-6782